4-Bromophthalimide 98%

Supplier: Ambeed
Warning

A292220-100G A292220-10G A292220-1G A292220-25G A292220-5G
77194-348EA 122 USD
77194-348 77194-350 77194-352 77194-354 77194-356
4-Bromophthalimide 98%
4-Bromophthalimide

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MDL Number: MFCD00466049
CAS Number: 6941-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR