(R)-2-Amino-2-(4-chlorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A291886-100MG A291886-1G A291886-250MG A291886-50MG
77194-340EA 175.1 USD
77194-340 77194-342 77194-344 77194-346
(R)-2-Amino-2-(4-chlorophenyl)ethanol 95%
(R)-2-Amino-2-(4-chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD09253717
CAS Number: 1067658-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR