(4-methoxy-3-(morpholinosulfonyl)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A291428-1G A291428-250MG A291428-5G
77166-802EA 216 USD
77166-802 77166-804 77166-806
(4-methoxy-3-(morpholinosulfonyl)phenyl)boronic acid 95+%
(4-methoxy-3-(morpholinosulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₆S
MW: 301.12 g/mol
MDL Number: MFCD08056366
CAS Number: 1100095-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR