Boc-L-thyronine 97%

Supplier: Ambeed
Warning

A291369-100MG A291369-1G A291369-250MG
77194-320EA 88 USD
77194-320 77194-322 77194-324
Boc-L-thyronine 97%
Boc-L-thyronine

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₆
MDL Number: MFCD02682294
CAS Number: 1213135-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR