H-Orn(Boc)-OMe·HCl 95%

Supplier: Ambeed
Warning

Synonyms: H-Orn(Boc)-OMe.HCl, H-Orn(Boc)-OMe·HCl

A291346-100MG A291346-1G A291346-250MG A291346-25G
77166-794EA 5.15 USD
77166-794 77166-796 77166-798 77166-800
H-Orn(Boc)-OMe·HCl 95%
H-Orn(Boc)-OMe·HCl

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₄
MW: 282.765 g/mol
MDL Number: MFCD00237207
CAS Number: 2480-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR