N-Bromophthalimide 95%

Supplier: Ambeed
Warning

A291139-100G A291139-25G A291139-5G
77194-310EA 172 USD
77194-310 77194-312 77194-314
N-Bromophthalimide 95%
N-Bromophthalimide

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MDL Number: MFCD00005888
CAS Number: 2439-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR