SGI-1776 99+%

Supplier: Ambeed
Warning

Synonyms: N-((1-Methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine, SGI-1776 free base

A290934-10MG A290934-1G A290934-1MG A290934-50MG A290934-5MG A290934-100MG A290934-250MG
77194-300EA 21.63 USD
77194-300 77194-302 77194-304 77194-306 77194-308 77746-539 77746-540
SGI-1776 99+%
SGI-1776

Precaución: for Research Use Only

Formula: C₂₀H₂₂F₃N₅O
MW: 405.4 g/mol
MDL Number: MFCD16659064
CAS Number: 1025065-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR