1,2:7,8-Diepoxyoctane 98%

Supplier: Ambeed
Danger

Synonyms: 1,4-Di(oxiran-2-yl)butane

A290716-1G A290716-25G A290716-5G
77194-292EA 12 USD
77194-292 77194-294 77194-296
1,2:7,8-Diepoxyoctane 98%
1,2:7,8-Diepoxyoctane

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
MDL Number: MFCD00005155
CAS Number: 2426-07-5
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR