4-(4-Methoxybenzoyl)piperidine hydrochloride 97%

Supplier: Ambeed
Warning

A289311-1G A289311-250MG A289311-5G
77166-776EA 23 USD
77166-776 77166-778 77166-780
4-(4-Methoxybenzoyl)piperidine hydrochloride 97%
4-(4-Methoxybenzoyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClNO₂
MDL Number: MFCD00114817
CAS Number: 25519-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR