5-Chlorobenzofuran-2(3H)-one 97%

Supplier: Ambeed
Warning

A288652-100MG A288652-1G A288652-250MG A288652-5G
77166-768EA 36.05 USD
77166-768 77166-770 77166-772 77166-774
5-Chlorobenzofuran-2(3H)-one 97%
5-Chlorobenzofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClO₂
MW: 168.577 g/mol
MDL Number: MFCD09842447
CAS Number: 28033-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR