2-(5-Amino-1,3-dioxoisoindolin-2-yl)acetic acid 96%

Supplier: Ambeed
Warning

A288498-1G A288498-250MG A288498-5G
77194-264EA 8.24 USD
77194-264 77194-266 77194-268 77194-270
2-(5-Amino-1,3-dioxoisoindolin-2-yl)acetic acid 96%
2-(5-Amino-1,3-dioxoisoindolin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MDL Number: MFCD03163249
CAS Number: 10133-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR