Benzyl (5-aminobicyclo[3.2.1]octan-1-yl)carbamate 98%

Supplier: Ambeed
Warning

A288465-100MG A288465-1G A288465-250MG
77194-258EA 117.42 USD
77194-258 77194-260 77194-262
Benzyl (5-aminobicyclo[3.2.1]octan-1-yl)carbamate 98%
Benzyl (5-aminobicyclo[3.2.1]octan-1-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₂
MW: 274.35 g/mol
MDL Number: MFCD30185167
CAS Number: 1383973-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR