Thiazol-2-ylmethanamine 97%

Supplier: Ambeed
Danger

Synonyms: 2-(Aminomethyl)-1,3-thiazole, (1,3-Thiazol-2-yl)methylamine

A288400-100MG A288400-1G A288400-250MG
77166-756EA 11 USD
77166-756 77166-758 77166-760
Thiazol-2-ylmethanamine 97%
Thiazol-2-ylmethanamine

Precaución: for Research Use Only

Formula: C₄H₆N₂S
MDL Number: MFCD04113607
CAS Number: 55661-33-1
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR