4-Amino-6-(trifluoromethyl)benzene-1,3-disulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: DTA

A288221-100MG A288221-250MG A288221-1G A288221-5G
77194-248EA 8.24 USD
77194-248 77194-250 77616-238 77616-240
4-Amino-6-(trifluoromethyl)benzene-1,3-disulfonamide 95%
4-Amino-6-(trifluoromethyl)benzene-1,3-disulfonamide

Precaución: for Research Use Only

Formula: C₇H₈F₃N₃O₄S₂
MW: 319.28 g/mol
MDL Number: MFCD00197629
CAS Number: 654-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR