3-Chloro-2-methoxy-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A287136-1G A287136-250MG
77166-724EA 116 USD
77166-724 77166-726
3-Chloro-2-methoxy-4-(trifluoromethyl)pyridine 95%
3-Chloro-2-methoxy-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MDL Number: MFCD16610626
CAS Number: 1227563-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR