4-Chloro-3-(hydroxymethyl)phenol 95%

Supplier: Ambeed
Warning

A286974-1G A286974-250MG
77166-720EA 93.73 USD
77166-720 77166-722
4-Chloro-3-(hydroxymethyl)phenol 95%
4-Chloro-3-(hydroxymethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.582 g/mol
MDL Number: MFCD09038291
CAS Number: 876299-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR