2-Bromo-6-fluoropyridin-3-ol 98%

Supplier: Ambeed
Warning

A286561-1G A286561-250MG A286561-5G
77166-708EA 23 USD
77166-708 77166-710 77166-712
2-Bromo-6-fluoropyridin-3-ol 98%
2-Bromo-6-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MDL Number: MFCD18257625
CAS Number: 1256822-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR