Ethyl 4-chloro-6,7-difluoroquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A286333-1G A286333-250MG
77166-698EA 138.02 USD
77166-698 77166-700
Ethyl 4-chloro-6,7-difluoroquinoline-3-carboxylate 97%
Ethyl 4-chloro-6,7-difluoroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₈ClF₂NO₂
MW: 271.65 g/mol
MDL Number: MFCD06014061
CAS Number: 318685-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR