(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC

A285256-100MG A285256-1G A285256-1MG A285256-250MG A285256-5G
77194-216EA 69.01 USD
77194-216 77194-218 77194-220 77194-222 77194-224
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 97%
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₃₀N₄O₃S
MW: 430.56 g/mol
MDL Number: MFCD29049827
CAS Number: 1448297-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR