N-(4-Methoxybenzyl)oxetan-3-amine 97%

Supplier: Ambeed
Warning

A284837-100MG A284837-250MG
77194-210EA 79.31 USD
77194-210 77194-212
N-(4-Methoxybenzyl)oxetan-3-amine 97%
N-(4-Methoxybenzyl)oxetan-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.24 g/mol
MDL Number: MFCD18849180
CAS Number: 1344099-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR