1,3-Dichloro-2,4,6-trifluorobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-1,3,5-trifluorobenzene

A284679-1G A284679-250MG A284679-5G
77194-204EA 70.04 USD
77194-204 77194-206 77194-208
1,3-Dichloro-2,4,6-trifluorobenzene 97%
1,3-Dichloro-2,4,6-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆HCl₂F₃
MW: 200.97 g/mol
MDL Number: MFCD00012242
CAS Number: 2368-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR