2,3-Diamino-6-chloro-benzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diamino-6-chlorobenzonitrile

A284513-1G A284513-250MG
77166-672EA 29.87 USD
77166-672 77166-674
2,3-Diamino-6-chloro-benzonitrile 95%
2,3-Diamino-6-chloro-benzonitrile

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.596 g/mol
MDL Number: MFCD20723943
CAS Number: 548457-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR