3,3',3'',3'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakisphenol 95%

Supplier: Ambeed
Warning

A284334-100MG A284334-1G A284334-250MG A284334-5G
77166-664EA 22 USD
77166-664 77166-666 77166-668 77166-670
3,3',3'',3'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakisphenol 95%
3,3',3'',3'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakisphenol

Precaución: for Research Use Only

Formula: C₄₄H₃₀N₄O₄
MDL Number: MFCD06657911
CAS Number: 22112-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR