4-(Bromomethyl)-2-chlorobenzonitrile 95%

Supplier: Ambeed
Danger

A283944-100MG A283944-250MG
77194-200EA 20 USD
77194-200 77194-202
4-(Bromomethyl)-2-chlorobenzonitrile 95%
4-(Bromomethyl)-2-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrClN
MDL Number: MFCD18392461
CAS Number: 83311-25-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR