(2-(Azetidin-1-yl)pyrimidin-5-yl)boronic acid 96%

Supplier: Ambeed
Warning

A283017-1G A283017-250MG
77166-652EA 248.23 USD
77166-652 77166-654
(2-(Azetidin-1-yl)pyrimidin-5-yl)boronic acid 96%
(2-(Azetidin-1-yl)pyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BN₃O₂
MW: 178.98 g/mol
MDL Number: MFCD18261197
CAS Number: 1411643-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR