1,7-Diaminoheptane 98%

Supplier: Ambeed
Danger

Synonyms: Heptamethylendiamine, Heptane-1,7-diamine, 1,7-Heptanediamine

A282922-1G A282922-25G A282922-5G
77194-192EA 12.36 USD
77194-192 77616-202 77746-517
1,7-Diaminoheptane 98%
1,7-Diaminoheptane

Precaución: for Research Use Only

Formula: C₇H₁₈N₂
MW: 130.23 g/mol
MDL Number: MFCD00008246
CAS Number: 646-19-5
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR