1,2'-Dinaphthylamine 98%

Supplier: Ambeed
Warning

A282694-100MG A282694-250MG
77194-188EA 13 USD
77194-188 77194-190
1,2'-Dinaphthylamine 98%
1,2'-Dinaphthylamine

Precaución: for Research Use Only

Formula: C₂₀H₁₅N
MDL Number: MFCD01632468
CAS Number: 4669-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR