1,2-Dioleoyl-sn-glycero-3-phosphocholine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2,3-Bis(oleoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, (R)-2,3-Bis(oleoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DOPC

A282380-1G A282380-5G A282380-100MG
77194-186EA 168.92 USD
77194-186 77568-572 77746-512
1,2-Dioleoyl-sn-glycero-3-phosphocholine 98%
1,2-Dioleoyl-sn-glycero-3-phosphocholine

Precaución: For research use only.

Formula: C₄₄H₈₄NO₈P
MW: 786.13 g/mol
MDL Number: MFCD00135191
CAS Number: 4235-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR