1,14-Dibromo-3,6,9,12-tetraoxatetradecane 98%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG4-bromide

A282104-10G A282104-1G A282104-250MG A282104-5G A282104-100MG
77194-176EA 1109.31 USD
77194-176 77194-178 77194-180 77194-182 77746-509
1,14-Dibromo-3,6,9,12-tetraoxatetradecane 98%
1,14-Dibromo-3,6,9,12-tetraoxatetradecane

Precaución: for Research Use Only

Formula: C₁₀H₂₀Br₂O₄
MW: 364.071 g/mol
MDL Number: MFCD07369317
CAS Number: 57602-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR