1-(Methylamino)anthraquinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Methylamino)anthracene-9,10-dione

A281511-100G A281511-25G
77194-162EA 30.9 USD
77194-162 77194-164
1-(Methylamino)anthraquinone 98%
1-(Methylamino)anthraquinone

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₂
MW: 237.26 g/mol
MDL Number: MFCD00001197
CAS Number: 82-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR