tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate 95%
Supplier: Ambeed
|
Warning
|
A281254-100G
A281254-1G
A281254-25G
A281254-5G
77166-634EA
414
USD
77166-634
77166-636
77166-638
77166-640
77166-642
tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate 95%
tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₄H₂₇N₃O₃ |
MDL Number:
MFCD22190570 CAS Number: 365998-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...