(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one
A281246-100MG
A281246-1G
A281246-250MG
77194-142EA
21
USD
77194-142
77194-144
77194-146
(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one 97%
(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one
Precaución: for Research Use Only
Formula:
C₁₁H₁₁F₃N₂O MW: 244.22 g/mol |
MDL Number:
MFCD12401613 CAS Number: 1015078-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...