6-Phenoxypyridin-3-amine 98%

Supplier: Ambeed
Warning

A280767-1G A280767-5G
77166-630EA 27.81 USD
77166-630 77166-632
6-Phenoxypyridin-3-amine 98%
6-Phenoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.22 g/mol
MDL Number: MFCD01692449
CAS Number: 25194-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR