PF-06405761 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-N-(3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl)benzenesulphonamide, PFI-1

A280183-10MG A280183-1MG A280183-50MG A280183-5MG
77194-132EA 39 USD
77194-132 77194-134 77194-136 77194-138
PF-06405761 98%
PF-06405761

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₄S
MW: 347.4 g/mol
MDL Number: MFCD22580416
CAS Number: 1403764-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR