(R)-1-Cyclopropylethan-1-amine hydrochloride 97%

Supplier: Ambeed
Warning

A280016-100MG A280016-250MG A280016-1G
77194-128EA 14 USD
77194-128 77194-130 77616-188
(R)-1-Cyclopropylethan-1-amine hydrochloride 97%
(R)-1-Cyclopropylethan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN
MDL Number: MFCD19160583
CAS Number: 195252-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR