tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (5-oxotetrahydrofuran-3-yl)carbamate

A279537-100MG A279537-1G A279537-250MG
77194-122EA 6.18 USD
77194-122 77194-124 77194-126
tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate 97%
tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD04113636
CAS Number: 104227-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR