1-(2-Amino-6-methylphenyl)piperidin-2-one 97%

Supplier: Ambeed
Warning

A279086-100MG A279086-250MG
77194-116EA 50.47 USD
77194-116 77194-118
1-(2-Amino-6-methylphenyl)piperidin-2-one 97%
1-(2-Amino-6-methylphenyl)piperidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
MDL Number: MFCD13705089
CAS Number: 1211496-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR