2-Bromo-5-chloro-4-fluoropyridine 97%

Supplier: Ambeed
Warning

A278954-100MG A278954-1G A278954-250MG
77194-110EA 80.34 USD
77194-110 77194-112 77194-114
2-Bromo-5-chloro-4-fluoropyridine 97%
2-Bromo-5-chloro-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
MDL Number: MFCD11100659
CAS Number: 1033203-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR