tert-Butyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A278839-1G A278839-250MG A278839-5G
77194-104EA 45 USD
77194-104 77194-106 77194-108
tert-Butyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate 97%
tert-Butyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD18831466
CAS Number: 197229-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR