2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2-Dimethyl-3-propyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide, 1,2-Dimethyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide, [DMPIIm]
A278658-1G
A278658-250MG
A278658-25G
A278658-5G
77194-096EA
11
USD
77194-096
77194-098
77194-100
77194-102
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide 98%
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide
Precaución: for Research Use Only
Formula:
C₁₀H₁₅F₆N₃O₄S₂ |
MDL Number:
MFCD03788922 CAS Number: 169051-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...