1-tert-Butyl 3-ethyl 2-oxopyrrolidine-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

A278619-100MG A278619-1G A278619-250MG A278619-5G
77166-600EA 22.66 USD
77166-600 77166-602 77166-604 77166-606
1-tert-Butyl 3-ethyl 2-oxopyrrolidine-1,3-dicarboxylate 95%
1-tert-Butyl 3-ethyl 2-oxopyrrolidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.28 g/mol
MDL Number: MFCD24466252
CAS Number: 188528-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR