1,2-Dichloro-4-methoxy-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A278305-250MG A278305-1G
77194-094EA 88.58 USD
77194-094 77746-499
1,2-Dichloro-4-methoxy-3-nitrobenzene 97%
1,2-Dichloro-4-methoxy-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₃
MW: 222.02 g/mol
MDL Number: MFCD28787075
CAS Number: 1806275-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR