1,3-Bis(3-aminophenoxy)benzene 97%

Supplier: Ambeed
Warning

A277796-10G A277796-1G A277796-5G A277796-100G A277796-25G
77194-088EA 82.4 USD
77194-088 77194-090 77194-092 77550-706 77550-708
1,3-Bis(3-aminophenoxy)benzene 97%
1,3-Bis(3-aminophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₂
MW: 292.3318 g/mol
MDL Number: MFCD00043718
CAS Number: 10526-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR