Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

A277581-1G A277581-250MG A277581-5G
77166-578EA 43.26 USD
77166-578 77166-580 77166-582
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
MDL Number: MFCD02187526
CAS Number: 70705-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR