(S)-3-Amino-3-(2-methylphenyl)propionic acid 98%

Supplier: Ambeed
Warning

A277455-10G A277455-1G A277455-250MG A277455-5G
77194-078EA 242 USD
77194-078 77194-080 77194-082 77194-084
(S)-3-Amino-3-(2-methylphenyl)propionic acid 98%
(S)-3-Amino-3-(2-methylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD04113679
CAS Number: 736131-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR