1-(2-Aminothiazol-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A276872-100MG A276872-250MG A276872-5G A276872-1G
77194-066EA 15 USD
77194-066 77194-068 77194-070 77746-494
1-(2-Aminothiazol-4-yl)ethanone 97%
1-(2-Aminothiazol-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MDL Number: MFCD06090850
CAS Number: 101258-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR