1-(Phenylsulfonyl)-1H-indole-3-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Phenylsulphonyl)-1H-indole-3-carbaldehyde, 3-Formyl-1-(phenylsulphonyl)-1H-indole

A276671-100MG A276671-1G A276671-250MG A276671-5G
77194-058EA 24.72 USD
77194-058 77194-060 77194-062 77194-064
1-(Phenylsulfonyl)-1H-indole-3-carboxaldehyde 97%
1-(Phenylsulfonyl)-1H-indole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₃S
MW: 285.318 g/mol
MDL Number: MFCD02681985
CAS Number: 80360-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR