4-Bromo-2-fluoro-6-methylbenzonitrile 95%

Supplier: Ambeed
Warning

A276004-100MG A276004-250MG
77166-562EA 41.2 USD
77166-562 77166-564
4-Bromo-2-fluoro-6-methylbenzonitrile 95%
4-Bromo-2-fluoro-6-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.034 g/mol
MDL Number: MFCD21337355
CAS Number: 1427438-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR