tert-Butyl 7-amino-2-azaspiro[3.5]nonane-2-carboxylate 98%

Supplier: Ambeed
Warning

A275983-1G A275983-250MG A275983-100MG
77166-558EA 808.55 USD
77166-558 77166-560 77683-836
tert-Butyl 7-amino-2-azaspiro[3.5]nonane-2-carboxylate 98%
tert-Butyl 7-amino-2-azaspiro[3.5]nonane-2-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₄N₂O₂
MW: 240.34 g/mol
MDL Number: MFCD21648502
CAS Number: 1408075-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR