Boc-Lys-OtBu 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate

A275334-100MG A275334-1G A275334-250MG A275334-5G
77166-548EA 17.51 USD
77166-548 77166-550 77166-552 77166-554
Boc-Lys-OtBu 98%
Boc-Lys-OtBu

Precaución: for Research Use Only

Formula: C₁₅H₃₀N₂O₄
MW: 302.4 g/mol
MDL Number: MFCD28963513
CAS Number: 7750-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR